missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10632515
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632515 250 mg Amber Glass Bottle 250mg
10215674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10632515 Supplier Thermo Scientific Maybridge Supplier No. CC52404CB
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 215460-40-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD09064964
InChI Key SIHCMIVOQNBOFV-UHFFFAOYSA-N
Synonym 4-tetrahydropyran-4-yloxy benzaldehyde, 4-oxan-4-yloxy benzaldehyde, 4-tetrahydro-2h-pyran-4-yloxy benzaldehyde, 4-4-tetrahydropyranyloxy benzaldehyde, benzaldehyde,4-tetrahydro-2h-pyran-4-yl oxy, 4-tetrahydro-2h-pyran-4-yl oxy benzaldehyde, 4-oxan-4-yl oxy benzaldehyde, 4-4-formylphenoxy tetrahydro-2h-pyran, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzaldehyde
PubChem CID 15034751
IUPAC Name 4-(oxan-4-yloxy)benzaldehyde
SMILES O=CC1=CC=C(OC2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 4-(Tetrahydropyran-4-yloxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.