missing translation for 'onlineSavingsMsg'
Learn More

(4-Thien-2-yltetrahydropyran-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10367734
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367734 250 mg Amber Glass Bottle 250mg
10586334 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10367734 Supplier Thermo Scientific Maybridge Supplier No. CC62209CB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-94-1
Molecular Formula C10H14O2S
Molecular Weight (g/mol) 198.28
MDL Number MFCD09879932
InChI Key JHVRUEHWCGLJPP-UHFFFAOYSA-N
Synonym 4-thien-2-yltetrahydropyran-4-yl methanol, 4-thiophen-2-yl oxan-4-yl methanol, 4-thiophen-2-yltetrahydropyran-4-yl methanol, 4-thien-2-yltetrahydro-2h-pyran-4-yl methanol, 4-2-thienyl tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl-4-thien-2-yltetrahydro-2h-pyran, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methanol, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-yl methan-1-ol
PubChem CID 24229668
IUPAC Name [4-(thiophen-2-yl)oxan-4-yl]methanol
SMILES OCC1(CCOCC1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 198.28
Percent Purity 97%
Chemical Name or Material (4-Thien-2-yltetrahydropyran-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.