Learn More
(4-Thien-3-ylphenyl)methanol, ≥97%, Maybridge™
Brand: Maybridge CC48709DE
Code : 31
Additional Details : CAS Number : 160278-20-6 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
160278-20-6 | |
190.26 | |
KEHJEXSFOWCTHM-UHFFFAOYSA-N | |
15186379 | |
C1=CC(=CC=C1CO)C2=CSC=C2 |
C11H10OS | |
MFCD06203084 | |
4-thien-3-ylphenyl methanol, 4-thiophen-3-yl phenyl methanol, benzenemethanol, 4-3-thienyl, 4-3-thienyl phenyl methanol, 4-thien-3-yl-benzyl alcohol, benzenemethanol,4-3-thienyl, 4-3-thienyl-phenyl-methanol, 4-3-thienyl phenyl methan-1-ol | |
(4-thiophen-3-ylphenyl)methanol |
Specifications
(4-Thien-3-ylphenyl)methanol | |
160278-20-6 | |
100.0 | |
C11H10OS | |
4-thien-3-ylphenyl methanol, 4-thiophen-3-yl phenyl methanol, benzenemethanol, 4-3-thienyl, 4-3-thienyl phenyl methanol, 4-thien-3-yl-benzyl alcohol, benzenemethanol,4-3-thienyl, 4-3-thienyl-phenyl-methanol, 4-3-thienyl phenyl methan-1-ol | |
C1=CC(=CC=C1CO)C2=CSC=C2 | |
190.26 | |
190.27 |
5g | |
96.5 | |
97% | |
MFCD06203084 | |
KEHJEXSFOWCTHM-UHFFFAOYSA-N | |
(4-thiophen-3-ylphenyl)methanol | |
15186379 | |
97% |