missing translation for 'onlineSavingsMsg'
Learn More

4-Thiouracil, TRC

Product Code. 30012899
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012899 1 g 1g
30008076 5 g 5g
30009372 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012899 Supplier Toronto Research Chemicals Supplier No. TRCT3754051G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-28-6
Molecular Formula C4 H4 N2 O S
Molecular Weight (g/mol) 128.152
Synonym 3,4-Dihydro-4-thioxo-2(1H)-pyrimidinone, 2-Hydroxy-4-mercaptopyrimidine, 4-Thiopyrimidin-2-one, NSC 43288;
IUPAC Name 4-sulfanylpyrimidin-2-ol
SMILES Oc1nccc(S)n1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H4N2OS/c7-4-5-2-1-3(8)6-4/h1-2H,(H2,5,6,7,8)
Formula Weight 128.004
Percent Purity >95%
Chemical Name or Material 4-Thiouracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.