missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4-Thiouridine

Product Code. 18749776
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749776 Supplier Tocris Bioscience™ Supplier No. 7005/25
  • ONLINE EXCLUSIVE PRICE
    236.00 €
    222.60 € / 25mg
    Save 13.40 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Photoreactive uridine analog for RNA sequencing

4-Thiouridine is a photoreactive uridine analog for RNA sequencing (RNA seq) and thiol(SH)-linked alkylation for the metabolic sequencing of RNA (SLAM seq). Used as a crosslinker to identify RNA binding proteins. Is incorporated into newly transcribed RNA and can be used to track RNA from synthesis to degradation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Thiouridine
CAS 13957-31-8
Quantity 25 mg
Molecular Formula C9H12N2O5S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.