missing translation for 'onlineSavingsMsg'
Learn More

4-Tolylboronic acid, 97%

Product Code. 10693712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10693712 5 g Glass bottle 5g
10536841 1 g Glass bottle 1g
10016263 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10693712 Supplier Thermo Scientific Acros Supplier No. 344430050
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5720-05-8 | C7H9BO2 | 135.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-05-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00039138
InChI Key BIWQNIMLAISTBV-UHFFFAOYSA-N
Synonym 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
PubChem CID 79799
IUPAC Name (4-methylphenyl)boronic acid
SMILES CC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 256°C to 263°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C6H4B(OH)2
Quantity 5 g
Formula Weight 135.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Tolylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Tolylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.