missing translation for 'onlineSavingsMsg'
Learn More

4-Tolylhydrazine Monohydrochloride, TRC

Product Code. 30453862
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453862 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453862 Supplier Toronto Research Chemicals Supplier No. TRCT53661025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Tolylhydrazine Monohydrochloride
CAS 637-60-5
Molecular Formula C7H11ClN2
Quantity 25 g
Synonym (4-Methylphenyl)-hydrazine Hydrochloride, (4-Methylphenyl)-hydrazine Monohydrochloride, p-Tolyl-Hydrazine Hydrochloride, p-Tolyl-Hydrazine Monohydrochloride, N-(4-Methylphenyl)hydrazine Hydrochloride, p-Methylphenylhydrazine GHydrochloride, p-Tolylhydrazine Hydrochloride
InChI Formula InChI=1S/C7H10N2.ClH/c1-6-2-4-7(9-8)5-3-6;/h2-5,9H,8H2,1H3;1H
SMILES Cl.Cc1ccc(NN)cc1
IUPAC Name (4-methylphenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 158.63
Formula Weight 158.0611
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.