missing translation for 'onlineSavingsMsg'
Learn More

4' '-trans-Hydroxy Cilostazol, TRC

Product Code. 30136065
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136065 25 mg 25mg
30138199 50 mg 50mg
30005213 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136065 Supplier Toronto Research Chemicals Supplier No. TRCH91830025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87153-04-6
Molecular Formula C20 H27 N5 O3
Molecular Weight (g/mol) 385.46
Synonym 4' '-Trans-Hydroxy Cilostazol
IUPAC Name 6-[4-[1-(4-hydroxycyclohexyl)tetrazol-5-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES O[C@@H]1CC[C@H](CC1)n2nnnc2CCCCOc3ccc4NC(=O)CCc4c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H27N5O3/c26-16-7-5-15(6-8-16)25-19(22-23-24-25)3-1-2-12-28-17-9-10-18-14(13-17)4-11-20(27)21-18/h9-10,13,15-16,26H,1-8,11-12H2,(H,21,27)/t15-,16-
Formula Weight 385.2114
Chemical Name or Material 4' '-trans-Hydroxy Cilostazol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.