missing translation for 'onlineSavingsMsg'
Learn More

4-(Tri-n-butylstannyl)pyridine, 96%

Product Code. 15448087
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448087 1 g 1g
15458087 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448087 Supplier Thermo Scientific Alfa Aesar Supplier No. H51435.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124252-41-1 | C17H31NSn | 368.15 g/mol

4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

Solubility
Not miscible in water. Soluble in ether, hexane and tetrahydrofuran.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124252-41-1
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.15
MDL Number MFCD01319071
InChI Key UNEPXPMBVGDXGH-UHFFFAOYSA-N
PubChem CID 2737318
IUPAC Name tributyl(pyridin-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=NC=C1

Specifications

Quantity 1 g
UN Number UN2788
Synonym 4-tributylstannyl pyridine, pyridine,4-tributylstannyl, 4-tributylstannylpyridine, 4-tributylstannyl-pyridine, 4-tri-n-butylstannyl pyridine, pyridin-4-yl tributylstannane, pyridine, 4-tributylstannyl, 4-pyridyltributyltin, 4-pyridyl tributyltin, 4-tributyistannylpyridine
Solubility Information Not miscible in water. Soluble in ether,hexane and tetrahydrofuran.
Formula Weight 368.15
Percent Purity 96%
Chemical Name or Material 4-(Tri-n-butylstannyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.