missing translation for 'onlineSavingsMsg'
Learn More

4-(Tri-n-butylstannyl)thiazole, 95%

Product Code. 15418097
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418097 250 mg 250mg
15408097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418097 Supplier Thermo Scientific Alfa Aesar Supplier No. H51457.MD
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 173979-01-6 | C15H29NSSn | 374.17 g/mol

4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173979-01-6
Molecular Formula C15H29NSSn
Molecular Weight (g/mol) 374.17
MDL Number MFCD01319073
InChI Key YYQKQPYPLADFMK-UHFFFAOYSA-N
Synonym 4-tributylstannyl thiazole, 4-tributylstannyl-1,3-thiazole, 4-tributylstannylthiazole, 4-tri-n-butylstannyl thiazole, 4-tributylstannanyl-1,3-thiazole, 4-tributylstannanyl-thiazole, 4-tri-n-butylstannylthiazole, 4-thiazolyltri-n-butylstannane, tri-n-butyl-4-thiazolyltin
PubChem CID 2763258
SMILES CCCC[Sn](CCCC)(CCCC)C1=CSC=N1

Specifications

Quantity 250 mg
UN Number UN2788
Solubility Information Not miscible in water.
IUPAC Name tributyl(1,3-thiazol-4-yl)stannane
Formula Weight 374.17
Percent Purity 95%
Chemical Name or Material 4-(Tri-n-butylstannyl)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.