missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoroacetyl-p-phenylenediamine, 97%, Thermo Scientific™

Product Code. 10192854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192854 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192854 Supplier Thermo Scientific Acros Supplier No. 399945000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 53446-90-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 118°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H7F3N2O
Linear Formula NH2C6H4NHC(O)CF3
MDL Number MFCD00132978
Quantity 500 mg
Synonym 4-trifluoroacetamidoaniline, n-4-aminophenyl-2,2,2-trifluoroacetamide, 4-trifluoroacetamido aniline, 2,2,2-trifluoro-n-4-aminophenyl acetamide, 4-trifluoroacetylaminoaniline, 4-trifluoroacetyl amino aniline, 4-trifluoroacetyl-p-phenylenediamine, n1-trifluoroacetyl benzene-1,4-diamine, n-4-aminophenyl-2,2,2-tris fluoranyl ethanamide
InChI Key DEXJVEPWTWVUNM-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)NC(=O)C(F)(F)F
IUPAC Name N-(4-aminophenyl)-2,2,2-trifluoroacetamide
Molecular Weight (g/mol) 204.15
PubChem CID 675087
Formula Weight 204.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Trifluoroacetyl-p-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Trifluoroacetyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.