missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde, 99%

Product Code. 10551701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10551701 10 g Glass bottle 10g
10279253 1 g Glass bottle 1g
10549443 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10551701 Supplier Thermo Scientific Acros Supplier No. 293500100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 659-28-9 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 659-28-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00041530
InChI Key XQNVDQZWOBPLQZ-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
PubChem CID 69573
IUPAC Name 4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Green-Yellow
Density 1.3300g/mL
Boiling Point 80°C (16.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4CHO
Refractive Index 1.4570 to 1.459
Quantity 10 g
Specific Gravity 1.33
Formula Weight 190.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.