missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzeneboronic acid, 98%

Product Code. 11420847
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420847 1 g 1g
11430847 5 g 5g
11440847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420847 Supplier Thermo Scientific Alfa Aesar Supplier No. B23233.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 139301-27-2 | C7H6BF3O3 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139301-27-2
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01074648
InChI Key HUOFUOCSQCYFPW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxyphenylboronic acid, 4-trifluoromethoxy benzeneboronic acid, 4-trifluoromethoxy phenyl boronic acid, 4-trifluoromethoxy phenyl boranediol, p-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxy-phenyl boronic acid, boronic acid, 4-trifluoromethoxy phenyl, 4-trifluoromethoxy phenylboronicacid
PubChem CID 2734386
IUPAC Name [4-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OC(F)(F)F)C=C1

Specifications

Melting Point ∼120°C to 125°C
Quantity 1 g
Beilstein 8548201
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.