missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%

Product Code. 11442837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442837 25 g 25g
11422837 1 g 1g
11432837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442837 Supplier Thermo Scientific Alfa Aesar Supplier No. A15467.14
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 94108-56-2 | C7H4ClF3O3S | 260.611 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94108-56-2
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD00042408
InChI Key UHCDBMIOLNKDHG-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride
PubChem CID 523102
IUPAC Name 4-(trifluoromethoxy)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl

Specifications

Density 1.536
Boiling Point 120°C to 122°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.48
Quantity 25 g
UN Number UN3265
Beilstein 7641524
Sensitivity Moisture sensitive
Formula Weight 260.62
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.