missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isothiocyanate, 97%

Product Code. 15421029
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421029 1 g 1g
15431029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421029 Supplier Thermo Scientific Alfa Aesar Supplier No. H64013.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 64285-95-6 | C8H4F3NOS | 219.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64285-95-6
Molecular Formula C8H4F3NOS
Molecular Weight (g/mol) 219.181
MDL Number MFCD00042412
InChI Key JKSZUQPHKOPVHF-UHFFFAOYSA-N
PubChem CID 2777338
IUPAC Name 1-isothiocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=S)OC(F)(F)F

Specifications

Color Colorless
Density 1.35 g/mL
Boiling Point 215°C to 218°C (literature)
Flash Point 107°C (225°F)
Refractive Index 1.541
Quantity 1 g
UN Number UN1760
Beilstein 2723939
Sensitivity Moisture sensitive
Synonym 4-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethoxy benzene, benzene,1-isothiocyanato-4-trifluoromethoxy, benzene, 1-isothiocyanato-4-trifluoromethoxy, 4-trifluoromethoxy phenylisothiocyanate, 1-isothiocyanato-4-trifluoromethoxy-benzene, acmc-1b5bx, 4-trifluoromethoxyphenyl isothiocyanate, 4-trifluoromethoxyphenyl-isothiocyanate, 4-trifluoromethoxy benzenisothiocyanate
Formula Weight 219.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.