missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethoxyphenylboronic acid, 98%

Product Code. 10253042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10253042 1 g Glass bottle 1g
10519603 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10253042 Supplier Thermo Scientific Acros Supplier No. 359730010
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 139301-27-2 | C7H6BF3O3 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139301-27-2
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01074648
InChI Key HUOFUOCSQCYFPW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxyphenylboronic acid, 4-trifluoromethoxy benzeneboronic acid, 4-trifluoromethoxy phenyl boronic acid, 4-trifluoromethoxy phenyl boranediol, p-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxy-phenyl boronic acid, boronic acid, 4-trifluoromethoxy phenyl, 4-trifluoromethoxy phenylboronicacid
PubChem CID 2734386
IUPAC Name [4-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OC(F)(F)F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 123°C to 127°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 205.93
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Trifluoromethoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Trifluoromethoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.