missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1H-indazole, 97%, Thermo Scientific™

Product Code. 10785622
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10785622 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10785622 Supplier Thermo Scientific Maybridge Supplier No. MAY00293DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1000339-98-9
Assay Percent Range 97%
Molecular Formula C8H5F3N2
MDL Number MFCD09263211
Quantity 1 g
Synonym 4-trifluoromethyl-1h-indazole, indazole, 4-trifluoromethyl, 4-trifluoromethyl-1 2 h-indazole
InChI Key KCWIWQGNCLKDJZ-UHFFFAOYSA-N
SMILES FC(F)(F)C1=C2C=NNC2=CC=C1
IUPAC Name 4-(trifluoromethyl)-1H-indazole
Molecular Weight (g/mol) 186.14
PubChem CID 24729335
Formula Weight 186.14
Chemical Name or Material 4-(Trifluoromethyl)-1H-indazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.