missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-2(1H)-quinolinone, 97%, Thermo Scientific Chemicals

Product Code. 15408298
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408298 1 g 1g
15418298 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408298 Supplier Thermo Scientific Alfa Aesar Supplier No. H61732.03
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25199-84-2
Molecular Formula C10H6F3NO
Molecular Weight (g/mol) 213.159
MDL Number MFCD03407380
InChI Key UUROBWTVZZNDFD-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
PubChem CID 2759347
IUPAC Name 4-(trifluoromethyl)-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 213.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Trifluoromethyl-2(1H)-quinolinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.