missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-3-Pyridinecarboxylic Acid, TRC

Product Code. 17992105
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992105 1 g 1g
17931565 100 mg 100mg
17981775 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992105 Supplier Toronto Research Chemicals Supplier No. TRCT7920651G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158063-66-2
Molecular Formula C7 H4 F3 N O2
Molecular Weight (g/mol) 191.11
Synonym 4-(Trifluoromethyl)-3-pyridinecarboxylic acid, 4-Trifluoromethylnicotinic acid, 4-Trifluoromethylpyridine-3-carboxylic acid, TFNA
IUPAC Name 4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)c1cnccc1C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H4F3NO2/c8-7(9,10)5-1-2-11-3-4(5)6(12)13/h1-3H,(H,12,13)
Formula Weight 191.0194
Chemical Name or Material 4-(Trifluoromethyl)-3-Pyridinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.