missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzamide, 98%, BLD Pharm™

Product Code. 30793005
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30793005 10 g 10g
30786050 25 g 25g
30795879 500 g -
30790047 5 g 5g
30786601 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30793005 Supplier BLD Pharm Supplier No. BD1096010g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1891-90-3
Molecular Formula C8H6F3NO
MDL Number MFCD00007998
Quantity 10 g
SMILES NC(=O)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 189.14
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.