missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 15595196 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15595196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15595196 Supplier Thermo Scientific Acros Supplier No. 456560010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 158063-66-2
Melting Point 136.0°C to 141.0°C
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C7H4F3NO2
MDL Number MFCD00082626
Quantity 1 g
Synonym 4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
InChI Key LMRJHNFECNKDKH-UHFFFAOYSA-N
SMILES OC(=O)C1=C(C=CN=C1)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol) 191.11
PubChem CID 2777549
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)nicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)nicotinic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.