missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinic acid, 98%

Product Code. 11378946
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378946 250 mg 250mg
11368946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378946 Supplier Thermo Scientific Alfa Aesar Supplier No. L19553.MD
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 158063-66-2 | C7H4F3NO2 | 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158063-66-2
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD00082626
InChI Key LMRJHNFECNKDKH-UHFFFAOYSA-N
Synonym 4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
PubChem CID 2777549
IUPAC Name 4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(C=CN=C1)C(F)(F)F

Specifications

Melting Point 147°C to 149°C
Quantity 250 mg
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)nicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.