missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)diphenylacetylene, 99%

Product Code. 11461150
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461150 1 g 1g
11471150 5 g 5g
11481150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461150 Supplier Thermo Scientific Alfa Aesar Supplier No. H26300.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 136459-72-8 | C17H18Si | 250.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136459-72-8
Molecular Formula C17H18Si
Molecular Weight (g/mol) 250.416
MDL Number MFCD09953470
InChI Key UZRJWRFLYPENFH-UHFFFAOYSA-N
Synonym 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene
PubChem CID 21093331
IUPAC Name trimethyl-[4-(2-phenylethynyl)phenyl]silane
SMILES C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 37°C to 39°C
Quantity 1 g
Formula Weight 250.42
Percent Purity 99%
Chemical Name or Material 4-(Trimethylsilyl)diphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.