missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)morpholine, 97%

Product Code. 15481408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481408 25 g 25g
15461408 1 g 1g
15471408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481408 Supplier Thermo Scientific Alfa Aesar Supplier No. H55229.14
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13368-42-8 | C7H17NOSi | 159.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13368-42-8
Molecular Formula C7H17NOSi
Molecular Weight (g/mol) 159.304
MDL Number MFCD00010098
InChI Key JJOWIQMPCCUIGA-UHFFFAOYSA-N
Synonym 4-trimethylsilyl morpholine, n-trimethylsilyl morpholine, n-trimethylsilylmorpholine, trimethyl morpholin-4-yl silane, morpholine, 4-trimethylsilyl, trimethylsilyl morpholin, morpholinotrimethylsilane, trimethylmorpholinosilane, acmc-1btin, timtec-bb sbb009054
PubChem CID 559763
IUPAC Name trimethyl(morpholin-4-yl)silane
SMILES C[Si](C)(C)N1CCOCC1

Specifications

Density 0.897
Boiling Point 160°C
Flash Point 29°C (84°F)
Refractive Index 1.441
Quantity 25 g
UN Number UN2924
Beilstein 1071480
Formula Weight 159.3
Percent Purity 97%
Chemical Name or Material 4-(Trimethylsilyl)morpholine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.