missing translation for 'onlineSavingsMsg'
Learn More

4-Tritylaniline, 97%

Product Code. 11468833
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468833 25 g 25g
11458833 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468833 Supplier Thermo Scientific Alfa Aesar Supplier No. A10550.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 22948-06-7 | C25H21N | 335.45 g/mol

4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22948-06-7
Molecular Formula C25H21N
Molecular Weight (g/mol) 335.45
MDL Number MFCD00007898
InChI Key XYHDHXBLSLSXSR-UHFFFAOYSA-N
Synonym p-tritylaniline, benzenamine, 4-triphenylmethyl, 4-triphenylmethyl aniline, p-aminotetraphenylmethane, 4-aminotetraphenylmethane, triphenyl p-aminophenyl methane, 4-trityl aniline, 4-triphenylmethyl benzenamine, p-toluidine, alpha,alpha,alpha-triphenyl, 4-aminophenyl triphenylmethane
PubChem CID 89914
IUPAC Name 4-tritylaniline
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)N

Specifications

Melting Point 256°C to 260°C
Quantity 25 g
Beilstein 2750328
Solubility Information Insoluble in water.
Formula Weight 335.45
Percent Purity 97%
Chemical Name or Material 4-Tritylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.