Learn More
TubeWriter 400 - Standard Rack Side Print for 0.5-2 mL Cryogenic Vials and Screw Cap Tubes
Product Code. 30067407
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
Product Code. Product Type Unit Size
30067407 Labeling Rack Each
1 options
Product Code. 30067407 Supplier TubeWriter Supplier No. PS1800CVEL120400
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Standard rack for directly printing on the side of 0.5 to 2 mL cryogenic vials and screw cap tubes
  • Standard printing rack designed for the 400
  • Prints and labels directly on the side of 0.5 to 2 mL cryogenic vials and screw cap tubes
  • Rack ensures correct alignment and holds vials securely
  • No time-consuming sticky labels or writing required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -807°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Product Type Labeling Rack