Learn More
Testo™ 435-4 Multi-function IAQ/HVAC Meter with Memory & Differential Pressure
Product Code. 10017792
Change view
Click to view available options
Airflow Rate:
0 to 3937 FPM
Unit Size:
Each
Product Code. Airflow Rate unitSize
10017792 0 to 3937 FPM Each
1 options
Product Code. 10017792 Supplier Testo™ Supplier No. 0563 4354
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Carry out all measurements that are necessary for checking and adjusting ventilation and A/C systems and for indoor air quality testing. Includes pressure difference sensor for pitot static tube or filter monitoring.
  • The extremely versatile Testo 435-4 can measure a host of variables including air temperature, air flow, volumetric flow rate, humidity and lux
  • You can configure it using any one of the many optional sensors and probes
  • You can also connect up to 3 wireless sensors whose measurements can be picked up by a wireless module (optional) over a distance of up to 65 feet
  • This allows you to carry out quick and easy measurements at places which would otherwise be extremely difficult to reach
  • There's also a special optional carry case that can hold both your meter and the probes and sensors that make up your individual set
  • Multifunction meter for A/C, ventilation and IAQ with memory
  • Accepts a wide array of sensors / probes for: airflow, temperature, humidity, CO2 and CO measurement
  • Performs timed and multi-point averaging and CFM, BTUh, and cooling tons calculation
  • Internal precision pressure sensor available
  • Printing capabilities (measurement, with date and time)
  • Data logs up to 10,000 readings (optional)
  • Optional TopSafe protects from impact, dust and water
  • Protective transport case organizes instrument, probes and accessories
  • Takes readings from up to 3 wireless temperature / humidity probes
  • Integrated rubber boot and magnetic back
  • Measures air moisture, material equilibrium moisture, pressure dewpoint
  • Instrument memory for 10,000 readings
  • PC software for analysis, archiving and documenting data
  • Protection class IP54
Warranty
2 years
Specifications
Airflow Rate 0 to 3937 FPM
Case Material ABS, TPE, Metal
Gas Type Carbon Dioxide
Description IP54
Dimensions (L x W x H) 220 x 74 x 46 mm
Power Supply Details Alkali manganese, mignon, Type AA (200 hour battery life)
Weight (Metric) 428 g