missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ Offset Adapter, 45°
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Closure Size | GL14 |
| Closure Color | Black |
| Description | 45° for Exhaust Filter |
| Color | Black |
| Liner Material | HDPE |
| For Use With (Application) | Angled Connection |
| Material | HDPE |
| Product Type | Adapter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?