Learn More
SHOWA™ 451 Series Natural Rubber Palm Coated Thermal-Insulated Gloves
Product Code. 17777607
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17777607 Medium 10 pairs
17787607 Small 10 pairs
17767607 Large 10 pairs
17797607 X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17777607 Supplier SHOWA™ Supplier No. 45108M
  • ONLINE EXCLUSIVE PRICE
    57.70 €
    52.20 € / 10 pairs
    Save 5.50 €
    9% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Created with warmth in mind, the 451 thermal-insulated gloves are designed to be worn in cold weather.
Benifits
  • Thermal Insulation
  • Flexible
  • Breathable
  • Extra Grip
  • Seamless Knit
  • Skin-friendly
  • Abrasion-resistant
Features
  • Rough Grip
  • Ergonomic
  • Elasticated Wrist Cuff
  • Natural Rubber Palm Coating
Trades & Applications
  • Signalers
  • Brick Laying
  • Carpentry
  • Driving Machines
  • Masonry
  • Demolition
  • Shipping/Receiving
Specifications
Certifications/Compliance CE Cat II, EN 388:2016, EN 511:2006
Coated Area Palm
Construction Seamless Knit
Coating Material Natural Rubber
Color Gray
Material Cotton, Polyester
Size Medium
Liner Material Acrylic
Resistant To Abrasion Resistant
Series 451