Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Product Code. 17727617
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
5 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17727617 X-Large 5 pairs
17717617 Medium 5 pairs
17707617 Large 5 pairs
17737617 2X-Large 5 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17727617 Supplier SHOWA™ Supplier No. 47709XL
  • ONLINE EXCLUSIVE PRICE
    82.55 €
    79.45 € / 5 pairs
    Save 3.10 €
    3% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Designed to keep hands warm and dry in the most extreme environments, the 477 cold-resistant glove combines thermal insulation with tactical grip.
Benifits
  • Seamless knit
  • Cold Resistant
  • Water-resistant
  • Flexible
  • Abrasion-resistant
  • Extra Grip
  • Thermal insulation
  • Form-fitting
  • Impermeable
Features
  • Foam grip
  • Elasticated wrist cuff
  • Ergonomic
  • Foam nitrile palm dip
  • Insulated
  • Fully-coated nitrile
Trades & Applications
  • Concreting
  • Guttering and drain pipe
  • Insulation
  • Washing and cleaning
  • Planting & repotting
  • Bottling
  • Water/Sewage treatment
  • Plumbing
  • Drainage, piping
  • Shipping/Receiving
Specifications
Certifications/Compliance CE Cat II, EN 388:2016, EN 511:2006
Construction Seamless Knit
Coating Material Nitrile
Color Blue, Black
Material Nylon, Polyester
Size X-Large
Liner Material Acrylic
Resistant To Cold Resistant, Water Resistant, Abrasion Resistant
Series 477