missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BBP11 BBP12 R4900 Series Printer Ribbon

Product Code. 12712447 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12712447 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12712447 Supplier Brady™ Supplier No. 804793
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This ribbon has superb smear resistance at high temperatures and superior solvent resistance.

  • This ribbon has superb smear resistance at high temperatures and superior solvent resistance.

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) BBP11 Label Printer, BBP12 Label Printer
Material Polyester
Length (English) 76.55 yd.
Width (English) 2.56 in.
Length (Metric) 70 m
Width (Metric) 65 mm
Thickness (English) 0.00018 in.
Thickness (Metric) 0.0045 mm
Diameter (Metric) 12.7 mm I.D.
Dimensions (L x W) 70 m x 65 mm
Description Black 4900 Series Thermal Transfer Printer Ribbon
Printable Sides Ink side out
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.