missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 1000 Series Gastight™ Saltline Syringes

Product Code. 15192514
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15192514 5 mL Each
15102524 10 mL Each
15182514 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15192514 Supplier Hamilton™ Supplier No. 208336
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with solutions that have a high salt concentration

  • Equipped with inert and non-corrosive plunger
  • Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip

Applications:

For life science applications with salty buffers, mining applications with acids or trace metal analysis

Specifications

Autoclavable Not Autoclavable
Material Borosilicate Glass
Needle Gauge No Needle Included
Product Type Gastight Saltline Syringe
Volume (Metric) 5 mL
For Use With (Equipment) Microlab 600 Diluter Dispenser
Connection Type Bubble Free Prime (BFP)
Graduated Yes
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.