Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Brand: Hamilton™ 7637-01
Code : A0
Additional Details : Weight : 1.00000kg
Description
- ideal for dispensing volumes from 0.05 µL up to 5 µL
- The 7000 series is the only zero dead volume syringe
- The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
26s G | |
705 | |
50 μL |