Learn More
Kimberly-Clark Professional™ KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves
Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.
Brand: Kimberly-Clark Professional™ 38718
Additional Details : Weight : 2.41200kg
Description
- Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
- High degree of sensitivity and dexterity for inspecting for defects and imperfections
- Designed for critical quality control inspections
- Seamless knit nylon shell reduces skin irritation
- Low lint for cleanroom applications
- Silicone-free
Specifications
Seamless Knit | |
White | |
Nylon | |
Medium | |
Knit |
Uncoated | |
Ambidextrous | |
Inspection Glove | |
Automotive Assembly, Aerospace Engineering, and Quality Control Inspections | |
KleenGuard™ G35 |