missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 6 3500 D MWCO Standard RC Pre-wetted Dialysis Kits

Product Code. 15370732
Change view
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Unit Size:
1m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) unitSize
15370732 29 mm 6.4 mL/cm vol./length 1m
15360732 11.5 mm 1.1 mL/cm vol./length 1m
15380732 34 mm 9.3 mL/cm vol./length 1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15370732 Supplier Spectrum™ Supplier No. 132592T
  • ONLINE EXCLUSIVE PRICE
    193.00 €
    133.00 € / 1m
    Save 60.00 €
    31% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

MWCO 3.5 kDa
Diameter (Metric) 29 mm
Length (Metric) 6.4 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.