missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 6 2000 D MWCO Standard RC Pre-wetted Dialysis Kits
Click to view available options
Diameter (Metric):
11.5 mm
24 mm
29 mm
Length (Metric):
1.1 mL/cm vol./length
4.6 mL/cm vol./length
6.4 mL/cm vol./length
Unit Size:
1m
Description
Includes
- 1 meter of Spectra/Por 6 Dialysis Tubing
- 1 Standard Spectra/Por Closure (for buoyancy)
- 1 Weighted Spectra/Por Closure (for orientation)
- 5 Tubing Opening Picks
- Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
- Membrane packaged wet in preservative solution of 0.05% sodium azide
- Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
- Include a pair of closures and convenient tubing opening picks
Specifications
Specifications
| MWCO | 2 kDa |
| Diameter (Metric) | 11.5 mm |
| Length (Metric) | 1.1 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction