missing translation for 'onlineSavingsMsg'
Learn More

4a,5-Dihydro Lovastatin, TRC

Product Code. 30137629
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137629 5 mg 5mg
30009453 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137629 Supplier Toronto Research Chemicals Supplier No. TRCD4485705MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77517-29-4
Molecular Formula C24 H38 O5
Molecular Weight (g/mol) 406.56
Synonym Butanoic acid, 2-methyl-, (1S,3S,4aR,7S,8S,8aS)-1,2,3,4,4a,7,8,8a-octahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)-, Butanoic acid, 2-methyl-, 1,2,3,4,4a,7,8,8a-octahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α(R*),3α,4aα,7β,8β(2S*,4S*),8aβ]]-, (+)-Dihydromevinolin, Dihydrolovastatin, Dihydromevinolin, L 154883, MSD 883, Lovastatin RC A (USP)
IUPAC Name [(1S,3S,4aR,7S,8S,8aS)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,4,4a,7,8,8a-octahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C[C@@H]2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H38O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,14-21,23,25H,5,8-13H2,1-4H3/t14-,15-,16-,17-,18+,19+,20-,21-,23-/m0/s1
Formula Weight 406.2719
Chemical Name or Material Dihydro-lovastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.