missing translation for 'onlineSavingsMsg'
Learn More

4a-Hydroxy Ivermectin, TRC

Product Code. 30402151
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402151 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402151 Supplier Toronto Research Chemicals Supplier No. TRCH9997401MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4a-Hydroxy Ivermectin
CAS 86629-74-5
Molecular Formula C48 H74 O15
Quantity 1 mg
Synonym Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], avermectin A1a deriv.
InChI Formula InChI=1S/C48H74O15/c1-10-25(2)43-28(5)16-17-47(63-43)22-34-19-33(62-47)15-14-27(4)42(60-39-21-37(55-9)44(30(7)58-39)61-38-20-36(54-8)40(50)29(6)57-38)26(3)12-11-13-32-24-56-45-41(51)31(23-49)18-35(46(52)59-34)48(32,45)53/h11-14,18,25-26,28-30,33-45,49-51,53H,10,15-17,19-24H2,1-9H3/b12-11+,27-14+,32-13-/t25-,26+,28+,29+,30+,33-,34+,35+,36+,37+,38+,39+,40+,41-,42+,43-,44+,45-,47-,48-/m1/s1
SMILES CC[C@@H](C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C/6\CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)CO)O2
Molecular Weight (g/mol) 891.09
Formula Weight 890.5028
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.