missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 4B Chemical Protection Coveralls

Product Code. 17787719 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17787719 Large Pack of 40
17767719 Small Pack of 40
17777719 Medium Pack of 40
17797719 X-Large Pack of 40
17707729 2X-Large Pack of 40
17717729 3X-Large Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17787719 Supplier Uvex™ Supplier No. 9837511
  • ONLINE EXCLUSIVE PRICE
    426.00 €
    397.35 € / Pack of 40
    Save 28.65 €
    6% Off
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4B disposable coveralls that are safe and breathable at the same time, due to the microporous spray-tight spunbond polyethylene laminate.

  • Breathable, lightweight and flexible material for exceptional wearer comfort
  • Particle-tight and spray-tight
  • Free of silicone and paint wetting impairment substances
  • Material: Polypropylene non woven laminated with Polyethylene film
  • Orange taped seams for optimal protection and good visibility
  • Middle finger loops prevent sleeves sliding up the arm
  • Self-adhesive zipper flap and taped seams for optimal protection
  • Anti-static properties
  • Protection against infectious diseases
  • Skin-friendly non-woven material inside
  • Elasticated waistband for a perfect fit
  • Secure and convenient closures thanks to elasticated bands on hood, arms and legs
  • Two-way zipper
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White, Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Open
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance EN 14126, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 13034 - type 6B, EN 1149-5, EN 1073-2, DIN 32781
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.