missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 4B Chemical Protection Suit with Feet

Product Code. 17787639 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17787639 Medium Each
17777639 Small Each
17797639 Large Each
17707649 X-Large Each
17717649 2X-Large Each
17727649 3X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17787639 Supplier Uvex™ Supplier No. 8909410
  • ONLINE EXCLUSIVE PRICE
    12.60 €
    11.22 € / Pack of 1
    Save 1.38 €
    10% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Uvex 4B chemical protection suit is comfortable and breathable, while the spunbond lining guarantees a pleasantly soft fit.

  • PP spunbonded non-woven fabric with PE film
  • Breathable, lightweight and flexible material for exceptional wearer comfort
  • Orange taped seams for optimal protection and good visibility
  • Middle finger loops prevent the sleeves from riding up
  • Optimum fit for a high level of wearer comfort
  • Secure and comfortable fastenings thanks to elasticated bands on hood, arms and legs
  • Tailored boot sock for extra safety
  • Particle-tight and spray-tight
  • Anti-static properties
  • Protection against infectious diseases
  • Protection against pesticides
  • Free of silicone, plasticizers and substances that interfere with wetting agents
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White, Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Socks
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Chemical protection/PPE category III, EN 14605:2005+A1:2009 (type 4), EN ISO 13982-1:2004+A1:2010 (type 5) , EN 13034:2005+A1:2010 (type 6), EN 1149-5:2008, EN 14126:2003+AC:2004, EN 1073-2:2002, DIN 32781:2010
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.