missing translation for 'onlineSavingsMsg'
Learn More

4Beta-Phorbol 12-Myristate 13-Acetate, TRC

Product Code. 30004529
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004529 1 mg 1mg
30005498 5 mg 5mg
30007243 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004529 Supplier Toronto Research Chemicals Supplier No. TRCP3534751MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36 H56 O8
Molecular Weight (g/mol) 616.82
Synonym Myristic Acid 9-ester with (+)-1,1aα,1bβ,4,4a,7aα,7b,8,9,9a-Decahydro-4aβ,7bα,9β,9aα-tetrahydroxy-3-(hydroxymethyl)-1,1,6,8α-tetramethyl-5H-cyclopropa[3,4]benz[1,2-e]azulen-5-one 9a-Acetate, [1aR-(1aα,1bβ,4aβ,7aα,7bα,8α,9β,9aα)]-9a-(acetyloxy)-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1H-cyclopropa[3,4]benz[1,2-e]azulen-9-yl Ester Tetradecanoic Acid, 1H-Cyclopropa[3,4]benz[1,2-e]azulene Tetradecanoic Acid Deriv. 12-O-Tetradecanoylphorbol 13-Acetate, 12-Tetradecanoylphorbol 13-Acetate, 12-Tetradecanoylphorbol 13-Monoacetate, 13-O-Acetylphorbol 12-Myristate, 4β-Phorbol 12-myristate 13-Acetate, Factor A1, Factor A1 (croton oil) NSC 262244, PMA, PMA (tumor promoter), Phorbol 12-myristate 13-Acetate, Phorbol 12-tetradecanoate 13-Acetate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)[C@@H](C=C(CO)C[C@@]3(O)[C@H]2C=C(C)C3=O)C4C(C)(C)C14OC(=O)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30?,32-,34-,35-,36?/m1/s1
Formula Weight 616.3975
Chemical Name or Material 4Beta-Phorbol 12-Myristate 13-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.