missing translation for 'onlineSavingsMsg'
Learn More

(4S)-N-Crotonyl-4-isopropyl-2-oxazolidinone, 99%, Thermo Scientific™

Product Code. 10747602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10747602 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10747602 Supplier Thermo Scientific Acros Supplier No. 337790010
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 90719-29-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 55°C to 58°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H15NO3
MDL Number MFCD00075429
Quantity 1 g
Specific Rotation + 102.5
Synonym 4s-3-2e-but-2-enoyl-4-isopropyl-1,3-oxazolidin-2-one, hg^l@lanry gyifjffjamdp, 4s-n-crotonyl-4-isopropyl-2-oxazolidinone, 3-2-butenoyl-4beta-isopropyloxazolidine-2-one, 4s-3-crotonoyl-4beta-isopropyloxazolidine-2-one, s-+-3-crotonoyl-4-isopropyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-propan-2-yl-1,3-oxazolidin-2-one, 3-trans-3-methyl-2-propenoyl-4-s-isopropyl-2-oxazolidinone
InChI Key YZRHIMNEVFYTAJ-WTSVBCDHSA-N
SMILES CC=CC(=O)N1C(COC1=O)C(C)C
IUPAC Name (4S)-3-[(E)-but-2-enoyl]-4-propan-2-yl-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 197.23
PubChem CID 5702601
Formula Weight 197.23
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material (4S)-N-Crotonyl-4-isopropyl-2-oxazolidinone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.