missing translation for 'onlineSavingsMsg'
Learn More

5-[(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde, 97%, Thermo Scientific™

Product Code. 10690913
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10690913 1 g Amber Glass Bottle 1g
10485322 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10690913 Supplier Thermo Scientific Maybridge Supplier No. KM10754DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 116750-06-2
Molecular Formula C14H9NO4
Molecular Weight (g/mol) 255.229
MDL Number MFCD02180376
InChI Key ZRBXPAHJXKKXME-UHFFFAOYSA-N
Synonym 5-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl methyl-2-furaldehyde, 5-1,3-dioxoisoindol-2-yl methyl furan-2-carbaldehyde, n-5-formylfur-2-yl methyl phthalimide, 5-1,3-dioxo-1,3-diyhdro-2h-isoindol-2-yl methyl-2-furaldehyde, 5-1,3-dioxoisoindolin-2-yl methyl furan-2-carbaldehyde, 5-n-phthalimidomethyl furan-2-carboxaldehyde, 5-1,3-dioxobenzo c azolin-2-yl methyl furan-2-carbaldehyde, 5-1,3-dioxoisoindolin-2-yl methyl furan-2-carboxaldehyde, 5-1,3-dioxo-2,3-dihydro-1h-2-isoindolylmethyl-2-furaldehyde
PubChem CID 273878
IUPAC Name 5-[(1,3-dioxoisoindol-2-yl)methyl]furan-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC3=CC=C(O3)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 255.23
Percent Purity 97%
Chemical Name or Material 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.