missing translation for 'onlineSavingsMsg'
Learn More

5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole, 97%, Thermo Scientific™

Product Code. 10773033
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773033 250 mg 250mg
10089071 1 g 1g
10604955 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10773033 Supplier Thermo Scientific Maybridge Supplier No. AW00426CB
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306934-71-4
Molecular Formula C13H16N4S
Molecular Weight (g/mol) 260.36
MDL Number MFCD00729015
InChI Key IENGAZSTLAONAR-UHFFFAOYSA-N
Synonym 5-1,4-diazepan-1-yl-3-phenyl-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl-1,4-diazepane, 1h-1,4-diazepine,hexahydro-1-3-phenyl-1,2,4-thiadiazol-5-yl, maybridge1_000443, 1-3-phenyl-1,2,4-thiazol-5-yl homopiperazine, 5-1,4-diazaperhydroepinyl-3-phenyl-1,2,4-thiadiazole
PubChem CID 2737169
IUPAC Name 5-(1,4-diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
SMILES C1CNCCN(C1)C1=NC(=NS1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 260.36
Percent Purity 97%
Chemical Name or Material 5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.