missing translation for 'onlineSavingsMsg'
Learn More

[5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-2-thienyl]methanol, 90%, Thermo Scientific™

Product Code. 10693023
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10693023 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10693023 Supplier Thermo Scientific Maybridge Supplier No. MO00643DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 465514-19-6
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C10H9F3N2OS
MDL Number MFCD03407368
Quantity 1 g
Synonym 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thienyl methanol, 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophen-2-yl methanol, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophen-2-yl methanol, maybridge3_004455, 5-1-methyl-5-trifluoromethyl pyrazol-3-yl-2-thienyl methan-1-ol, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thienylmethanol
InChI Key RJIIPCWLMHMXQA-UHFFFAOYSA-N
SMILES CN1C(=CC(=N1)C2=CC=C(S2)CO)C(F)(F)F
IUPAC Name [5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophen-2-yl]methanol
Molecular Weight (g/mol) 262.25
PubChem CID 2780713
Formula Weight 262.25
Percent Purity 90%
Chemical Name or Material {5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-2-thienyl}methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.