missing translation for 'onlineSavingsMsg'
Learn More

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10641873
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10641873 1 g 1g
10406092 250 mg 250mg
10351995 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10641873 Supplier Thermo Scientific Maybridge Supplier No. KM05281DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-93-4
Molecular Formula C10H7F3N2OS
Molecular Weight (g/mol) 260.234
MDL Number MFCD00052163
InChI Key ITUYPRXQRPMRTL-UHFFFAOYSA-N
Synonym 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carbaldehyde, 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carbaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl-thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carbaldehyde, 2-thiophenecarboxaldehyde,5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarbaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarboxaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl thiophene-2-carbaldehyde
PubChem CID 2775667
IUPAC Name 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carbaldehyde
SMILES CN1C(=CC(=N1)C2=CC=C(S2)C=O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 260.24
Percent Purity 97%
Chemical Name or Material 5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.