missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Furyl)-1-methyl-1H-pyrazol-3-yl]methanol, ≥97%, Thermo Scientific™

Product Code. 10192844
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10192844 1 g Amber Glass Bottle 1g
10082304 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192844 Supplier Thermo Scientific Maybridge Supplier No. CC46909DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 876728-41-5
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.19
MDL Number MFCD08271948
InChI Key XKIFRVDOWHFUHG-UHFFFAOYSA-N
Synonym 5-2-furyl-1-methyl-1h-pyrazol-3-yl methanol, 5-fur-2-yl-1-methyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1-methylpyrazol-3-yl methanol, 5-furan-2-yl-1-methyl-1h-pyrazol-3-yl methanol, 5-fur-2-yl-3-hydroxymethyl-1-methyl-1h-pyrazole, 5-2-furyl-1-methylpyrazol-3-yl methan-1-ol, 5-2-furyl-3-hydroxymethyl-1-methyl-1h-pyrazole
PubChem CID 18525802
IUPAC Name [5-(furan-2-yl)-1-methylpyrazol-3-yl]methanol
SMILES CN1N=C(CO)C=C1C1=CC=CO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material [5-(2-Furyl)-1-methyl-1H-pyrazol-3-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.