missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)cyclohexane-1,3-dione, 97%, Thermo Scientific™

Product Code. 10506805
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10506805 5 g 5g
10121122 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506805 Supplier Thermo Scientific Maybridge Supplier No. CD11333DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1774-11-4
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00104796
InChI Key FYLTVHCMIYGVPZ-UHFFFAOYSA-N
Synonym 5-furan-2-yl cyclohexane-1,3-dione, 5-2-furyl cyclohexane-1,3-dione, 5-2-furyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-2-furanyl, 5-furan-2-yl-cyclohexane-1,3-dione, 1,3-cyclohexanedione,5-2-furanyl, 5-2-furanyl cyclohexane-1,3-dione, maybridge1_003103, 5-2-furyl cyclohexan-1,3-dione, 5-2-furyl-cyclohexane-1,3-dione
PubChem CID 699513
IUPAC Name 5-(furan-2-yl)cyclohexane-1,3-dione
SMILES C1C(CC(=O)CC1=O)C2=CC=CO2

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)cyclohexane-1, 3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.