missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10662885
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662885 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10662885 Supplier Thermo Scientific Maybridge Supplier No. CC47701CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 868755-62-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H6O3S
MDL Number MFCD05885497
Quantity 250 mg
Synonym 5-2-furyl thiophene-2-carboxylic acid, 5-furan-2-yl thiophene-2-carboxylic acid, 5-fur-2-ylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-2-furanyl
InChI Key ZASUJVQPFQRFPQ-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(S1)C1=CC=CO1
IUPAC Name 5-(furan-2-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol) 194.20
PubChem CID 2052621
Formula Weight 194.21
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)thiophene-2-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.