missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™

Product Code. 10381615
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10381615 25 g 25g
10049151 1 g 1g
10178254 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10381615 Supplier Thermo Scientific Maybridge Supplier No. CC18209EB
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
InChI Key CQCKNGVVCKWDLF-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methanol, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-4-thiazolyl thiophene-2-methanol, 5-2-methylthiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl thien-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methan-1-ol
PubChem CID 2776470
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(S2)CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 211.31
Percent Purity 95%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.